Anti-HRH1 Mouse Monoclonal Antibody [clone: 480054]

Supplier: R & D Systems
MAB4726
220021-000EA 561.11 USD
220021-000
Anti-HRH1 Mouse Monoclonal Antibody [clone: 480054]
Antibodies
Type: Primary
Antigen: HRH1
Clonality: Monoclonal
Clone: 480054
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR