Anti-TRAT1 Mouse Monoclonal Antibody [clone: 488806]

Supplier: R & D Systems
MAB4708
220020-996EA 561.11 USD
220020-996
Anti-TRAT1 Mouse Monoclonal Antibody [clone: 488806]
Antibodies
Type: Primary
Antigen: TRAT1
Clonality: Monoclonal
Clone: 488806
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR