Anti-FCRLB Mouse Monoclonal Antibody [clone: 454217]

Supplier: R & D Systems
MAB4705
220020-995EA 561.11 USD
220020-995
Anti-FCRLB Mouse Monoclonal Antibody [clone: 454217]
Antibodies
Type: Primary
Antigen Symbol: FCRLB
Antigen Name: Fc receptor-like B
Antigen Synonyms: FCRL2, FCRLM2, FCRLY, FREB-2, FcRY
Clonality: Monoclonal
Clone: 454217
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR