Anti-GRK7 Mouse Monoclonal Antibody [clone: 496831]

Supplier: R & D Systems
MAB4700
220020-992EA 561.11 USD
220020-992
Anti-GRK7 Mouse Monoclonal Antibody [clone: 496831]
Antibodies
Type: Primary
Antigen Symbol: GRK7
Antigen Name: G protein-coupled receptor kinase 7
Antigen Synonyms: GPRK7
Clonality: Monoclonal
Clone: 496831
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR