Anti-JAK3 Mouse Monoclonal Antibody [clone: 452524]

Supplier: R & D Systems
MAB46991
220020-990EA 594.37 USD
220020-990
Anti-JAK3 Mouse Monoclonal Antibody [clone: 452524]
Antibodies
Type: Primary
Antigen: JAK3
Clonality: Monoclonal
Clone: 452524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR