Anti-JAK3 Mouse Monoclonal Antibody [clone: 452506]

Supplier: R & D Systems
MAB4699
220020-989EA 577.02 USD
220020-989
Anti-JAK3 Mouse Monoclonal Antibody [clone: 452506]
Antibodies
Type: Primary
Antigen: JAK3
Clonality: Monoclonal
Clone: 452506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR