Anti-RAP2A/B Mouse Monoclonal Antibody [clone: 455410]

Supplier: R & D Systems
MAB4698
220020-988EA 577.02 USD
220020-988
Anti-RAP2A/B Mouse Monoclonal Antibody [clone: 455410]
Antibodies
Type: Primary
Antigen: RAP2A/B
Clonality: Monoclonal
Clone: 455410
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR