Anti-SEMA4A Mouse Monoclonal Antibody [clone: 741531]

Supplier: R & D Systems
MAB4694
102885-786EA 594.37 USD
102885-786
Anti-SEMA4A Mouse Monoclonal Antibody [clone: 741531]
Antibodies
Type: Primary
Antigen: SEMA4A
Clonality: Monoclonal
Clone: 741531
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR