Anti-FIGF Rat Monoclonal Antibody [clone: 90409]

Supplier: R & D Systems
MAB469
220020-985EA 763.57 USD
220020-985
Anti-FIGF Rat Monoclonal Antibody [clone: 90409]
Antibodies
Type: Primary
Antigen Symbol: FIGF
Antigen Name: c-fos induced growth factor
Antigen Synonyms: AI325264, VEGF-D, Vegfd
Clonality: Monoclonal
Clone: 90409
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR