Anti-NLRP2 Mouse Monoclonal Antibody [clone: 443625]

Supplier: R & D Systems
MAB4684
220020-984EA 594.37 USD
220020-984
Anti-NLRP2 Mouse Monoclonal Antibody [clone: 443625]
Antibodies
Type: Primary
Antigen: NLRP2
Clonality: Monoclonal
Clone: 443625
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR