Anti-IL6ST Rat Monoclonal Antibody [clone: 125604]

Supplier: R & D Systems
MAB468
220020-981EA 763.57 USD
220020-981
Anti-IL6ST Rat Monoclonal Antibody [clone: 125604]
Antibodies
Type: Primary
Antigen: IL6ST
Clonality: Monoclonal
Clone: 125604
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR