Anti-DFFA Mouse Monoclonal Antibody [clone: 495403]

Supplier: R & D Systems
MAB4679
220020-980EA 577.02 USD
220020-980
Anti-DFFA Mouse Monoclonal Antibody [clone: 495403]
Antibodies
Type: Primary
Antigen: DFFA
Clonality: Monoclonal
Clone: 495403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR