Anti-EPHB2 Rat Monoclonal Antibody [clone: 512001]

Supplier: R & D Systems
MAB4671
220020-975EA 561.11 USD
220020-975
Anti-EPHB2 Rat Monoclonal Antibody [clone: 512001]
Antibodies
Type: Primary
Antigen: EPHB2
Clonality: Monoclonal
Clone: 512001
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR