Anti-CD47 Mouse Monoclonal Antibody [clone: 472603]

Supplier: R & D Systems
MAB4670
220020-974EA 655.11 USD
220020-974
Anti-CD47 Mouse Monoclonal Antibody [clone: 472603]
Antibodies
Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: 472603
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR