Anti-EPHB2 Rat Monoclonal Antibody [clone: 512012]

Supplier: R & D Systems
MAB467
220020-973EA 594.37 USD
220020-973
Anti-EPHB2 Rat Monoclonal Antibody [clone: 512012]
Antibodies
Type: Primary
Antigen: EPHB2
Clonality: Monoclonal
Clone: 512012
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR