Anti-ITGB7 Mouse Monoclonal Antibody [clone: 473207]

Supplier: R & D Systems
MAB4669
220020-972EA 594.37 USD
220020-972
Anti-ITGB7 Mouse Monoclonal Antibody [clone: 473207]
Antibodies
Type: Primary
Antigen: ITGB7
Clonality: Monoclonal
Clone: 473207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR