Anti-NAALADL2 Mouse Monoclonal Antibody [clone: 817225]

Supplier: R & D Systems
MAB4665
102885-782EA 577.02 USD
102885-782
Anti-NAALADL2 Mouse Monoclonal Antibody [clone: 817225]
Antibodies
Type: Primary
Antigen: NAALADL2
Clonality: Monoclonal
Clone: 817225
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR