Anti-SLC27A2 Mouse Monoclonal Antibody [clone: 466106]

Supplier: R & D Systems
MAB4659
220020-970EA 566.89 USD
220020-970
Anti-SLC27A2 Mouse Monoclonal Antibody [clone: 466106]
Antibodies
Type: Primary
Antigen: SLC27A2
Clonality: Monoclonal
Clone: 466106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR