Anti-RSPO1 Mouse Monoclonal Antibody [clone: 422407]

Supplier: R & D Systems
MAB4658
220020-969EA 594.37 USD
220020-969
Anti-RSPO1 Mouse Monoclonal Antibody [clone: 422407]
Antibodies
Type: Primary
Antigen: RSPO1
Clonality: Monoclonal
Clone: 422407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR