Anti-LAMA1 Rat Monoclonal Antibody [clone: AL-4]

Supplier: R & D Systems
MAB4656
220020-968EA 616.06 USD
220020-968
Anti-LAMA1 Rat Monoclonal Antibody [clone: AL-4]
Antibodies
Type: Primary
Antigen: LAMA1
Clonality: Monoclonal
Clone: AL-4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR