Anti-PROKR1 Mouse Monoclonal Antibody [clone: 420849]

Supplier: R & D Systems
MAB4655
220020-967EA 577.02 USD
220020-967
Anti-PROKR1 Mouse Monoclonal Antibody [clone: 420849]
Antibodies
Type: Primary
Antigen: PROKR1
Clonality: Monoclonal
Clone: 420849
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR