Anti-PGF Rat Monoclonal Antibody [clone: 62503]

Supplier: R & D Systems
MAB4651
220020-966EA 763.57 USD
220020-966
Anti-PGF Rat Monoclonal Antibody [clone: 62503]
Antibodies
Type: Primary
Antigen: PGF
Clonality: Monoclonal
Clone: 62503
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR