Anti-TPT1 Mouse Monoclonal Antibody [clone: 488411]

Supplier: R & D Systems
MAB4648
220020-964EA 594.37 USD
220020-964
Anti-TPT1 Mouse Monoclonal Antibody [clone: 488411]
Antibodies
Type: Primary
Antigen: TPT1
Clonality: Monoclonal
Clone: 488411
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR