Anti-C9ORF3 Mouse Monoclonal Antibody [clone: 562912]

Supplier: R & D Systems
MAB4647
220020-963EA 561.11 USD
220020-963
Anti-C9ORF3 Mouse Monoclonal Antibody [clone: 562912]
Antibodies
Type: Primary
Antigen: C9ORF3
Clonality: Monoclonal
Clone: 562912
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR