Anti-GPR50 Mouse Monoclonal Antibody [clone: 461129]

Supplier: R & D Systems
MAB4645
220020-962EA 577.02 USD
220020-962
Anti-GPR50 Mouse Monoclonal Antibody [clone: 461129]
Antibodies
Type: Primary
Antigen Symbol: GPR50
Antigen Name: G protein-coupled receptor 50
Antigen Synonyms: H9, Mel1c
Clonality: Monoclonal
Clone: 461129
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR