Anti-GRAP2 Mouse Monoclonal Antibody [clone: 475804]

Supplier: R & D Systems
MAB4640
220020-960EA 594.37 USD
220020-960
Anti-GRAP2 Mouse Monoclonal Antibody [clone: 475804]
Antibodies
Type: Primary
Antigen Symbol: GRAP2
Antigen Name: GRB2-related adaptor protein 2
Antigen Synonyms: GADS, GRAP-2, GRB2L, GRBLG, GRID, GRPL, GrbX, Grf40, Mona, P38
Clonality: Monoclonal
Clone: 475804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR