Anti-SHH Rat Monoclonal Antibody [clone: 171001]

Supplier: R & D Systems
MAB464
220020-959EA 772.25 USD
220020-959
Anti-SHH Rat Monoclonal Antibody [clone: 171001]
Antibodies
Type: Primary
Antigen: SHH
Clonality: Monoclonal
Clone: 171001
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR