Anti-IL28A Rat Monoclonal Antibody [clone: 625616]

Supplier: R & D Systems
MAB4635
102885-780EA 772.25 USD
102885-780
Anti-IL28A Rat Monoclonal Antibody [clone: 625616]
Antibodies
Type: Primary
Antigen: IL28A
Clonality: Monoclonal
Clone: 625616
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR