Anti-SLAMF7 Rat Monoclonal Antibody [clone: 520914]

Supplier: R & D Systems
MAB46281
220020-957EA 577.02 USD
220020-957
Anti-SLAMF7 Rat Monoclonal Antibody [clone: 520914]
Antibodies
Type: Primary
Antigen: SLAMF7
Clonality: Monoclonal
Clone: 520914
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR