Anti-SLAMF7 Rat Monoclonal Antibody [clone: 520911]

Supplier: R & D Systems
MAB4628
220020-956EA 577.02 USD
220020-956
Anti-SLAMF7 Rat Monoclonal Antibody [clone: 520911]
Antibodies
Type: Primary
Antigen: SLAMF7
Clonality: Monoclonal
Clone: 520911
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR