Anti-STEAP4 Mouse Monoclonal Antibody [clone: 418714]

Supplier: R & D Systems
MAB4626
220020-955EA 577.02 USD
220020-955
Anti-STEAP4 Mouse Monoclonal Antibody [clone: 418714]
Antibodies
Type: Primary
Antigen: STEAP4
Clonality: Monoclonal
Clone: 418714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR