Anti-CD37 Mouse Monoclonal Antibody [clone: 424925]

Supplier: R & D Systems
MAB4625
220020-954EA 594.37 USD
220020-954
Anti-CD37 Mouse Monoclonal Antibody [clone: 424925]
Antibodies
Type: Primary
Antigen: CD37
Clonality: Monoclonal
Clone: 424925
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR