Anti-CLDN3 Mouse Monoclonal Antibody [clone: 385021]

Supplier: R & D Systems
MAB4620
220020-952EA 639.2 USD
220020-952
Anti-CLDN3 Mouse Monoclonal Antibody [clone: 385021]
Antibodies
Type: Primary
Antigen: CLDN3
Clonality: Monoclonal
Clone: 385021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR