Anti-TNFSF11 Rat Monoclonal Antibody [clone: 88227]

Supplier: R & D Systems
MAB462
220020-951EA 763.57 USD
220020-951
Anti-TNFSF11 Rat Monoclonal Antibody [clone: 88227]
Antibodies
Type: Primary
Antigen: TNFSF11
Clonality: Monoclonal
Clone: 88227
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR