Anti-CLDN17 Mouse Monoclonal Antibody [clone: 410509]

Supplier: R & D Systems
MAB4619
220020-950EA 577.02 USD
220020-950
Anti-CLDN17 Mouse Monoclonal Antibody [clone: 410509]
Antibodies
Type: Primary
Antigen: CLDN17
Clonality: Monoclonal
Clone: 410509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR