Anti-CLDN1 Rat Monoclonal Antibody [clone: 421203]

Supplier: R & D Systems
MAB4618
220020-949EA 594.37 USD
220020-949
Anti-CLDN1 Rat Monoclonal Antibody [clone: 421203]
Antibodies
Type: Primary
Antigen: CLDN1
Clonality: Monoclonal
Clone: 421203
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR