Anti-CD82 Mouse Monoclonal Antibody [clone: 423524]

Supplier: R & D Systems
MAB4616
220020-947EA 623.29 USD
220020-947
Anti-CD82 Mouse Monoclonal Antibody [clone: 423524]
Antibodies
Type: Primary
Antigen: CD82
Clonality: Monoclonal
Clone: 423524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR