Anti-CD81 Mouse Monoclonal Antibody [clone: 454720]

Supplier: R & D Systems
MAB4615
220020-946EA 623.29 USD
220020-946
Anti-CD81 Mouse Monoclonal Antibody [clone: 454720]
Antibodies
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 454720
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR