Anti-CALCR Mouse Monoclonal Antibody [clone: 462802]

Supplier: R & D Systems
MAB4614
220020-945EA 577.02 USD
220020-945
Anti-CALCR Mouse Monoclonal Antibody [clone: 462802]
Antibodies
Type: Primary
Antigen: CALCR
Clonality: Monoclonal
Clone: 462802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR