Anti-CD151 Rat Monoclonal Antibody [clone: 455807]

Supplier: R & D Systems
MAB4609
220020-944EA 594.37 USD
220020-944
Anti-CD151 Rat Monoclonal Antibody [clone: 455807]
Antibodies
Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 455807
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR