Anti-STAT3 Rat Monoclonal Antibody [clone: 805435]

Supplier: R & D Systems
MAB4607
102885-778EA 594.37 USD
102885-778
Anti-STAT3 Rat Monoclonal Antibody [clone: 805435]
Antibodies
Type: Primary
Antigen: STAT3
Clonality: Monoclonal
Clone: 805435
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR