Anti-REL Rat Monoclonal Antibody [clone: 321422]

Supplier: R & D Systems
MAB4606
220020-942EA 485.91 USD
220020-942
Anti-REL Rat Monoclonal Antibody [clone: 321422]
Antibodies
Type: Primary
Antigen: REL
Clonality: Monoclonal
Clone: 321422
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR