Anti-HNF4A Mouse Monoclonal Antibody [clone: 843716]

Supplier: R & D Systems
MAB4605
102885-776EA 639.2 USD
102885-776
Anti-HNF4A Mouse Monoclonal Antibody [clone: 843716]
Antibodies
Simple Western

Type: Primary
Antigen: HNF4A
Clonality: Monoclonal
Clone: 843716
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR