Anti-PRSS8 Mouse Monoclonal Antibody [clone: 530622]

Supplier: R & D Systems
MAB4599
102885-774EA 577.02 USD
102885-774
Anti-PRSS8 Mouse Monoclonal Antibody [clone: 530622]
Antibodies
Type: Primary
Antigen: PRSS8
Clonality: Monoclonal
Clone: 530622
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR