Anti-RICTOR Rat Monoclonal Antibody [clone: 454313]

Supplier: R & D Systems
MAB4598
220020-941EA 561.11 USD
220020-941
Anti-RICTOR Rat Monoclonal Antibody [clone: 454313]
Antibodies
Type: Primary
Antigen: RICTOR
Clonality: Monoclonal
Clone: 454313
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR