Anti-TNFRSF11B Rat Monoclonal Antibody [clone: 110511]

Supplier: R & D Systems
MAB4591
220020-939EA 772.25 USD
220020-939
Anti-TNFRSF11B Rat Monoclonal Antibody [clone: 110511]
Antibodies
Type: Primary
Antigen: TNFRSF11B
Clonality: Monoclonal
Clone: 110511
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR