Anti-ROCK1 Mouse Monoclonal Antibody [clone: 701118]

Supplier: R & D Systems
MAB4590
102885-772EA 594.37 USD
102885-772
Anti-ROCK1 Mouse Monoclonal Antibody [clone: 701118]
Antibodies
Type: Primary
Antigen: ROCK1
Clonality: Monoclonal
Clone: 701118
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR