Anti-TNFRSF11B Rat Monoclonal Antibody [clone: 110517]

Supplier: R & D Systems
MAB459
220020-938EA 763.57 USD
220020-938
Anti-TNFRSF11B Rat Monoclonal Antibody [clone: 110517]
Antibodies
Type: Primary
Antigen: TNFRSF11B
Clonality: Monoclonal
Clone: 110517
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR