Anti-MAP3K8 Mouse Monoclonal Antibody [clone: 634130]

Supplier: R & D Systems
MAB4586
102885-770EA 594.37 USD
102885-770
Anti-MAP3K8 Mouse Monoclonal Antibody [clone: 634130]
Antibodies
Type: Primary
Antigen: MAP3K8
Clonality: Monoclonal
Clone: 634130
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR