Anti-EFNA2 Rat Monoclonal Antibody [clone: 84019]

Supplier: R & D Systems
MAB458
220020-937EA 763.57 USD
220020-937
Anti-EFNA2 Rat Monoclonal Antibody [clone: 84019]
Antibodies
Type: Primary
Antigen: EFNA2
Clonality: Monoclonal
Clone: 84019
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR